Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

Qnuclear™ Deep Red Stain

Catalog number: Q10363

Molecular Probes™  Related applications: Cell Structure | Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Qnuclear™ Deep Red stain is a highly selective nuclear counterstain designed for use with cells labeled with Qdot® 525, 565, 585, 605 and 625 nanocrystals and mounted in Qmount™ Qdot® Mounting Media (Q10336).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Color: Red, Deep Red
Label or Dye: Qnuclear™ Deep Red stain
Product Size: 100 µL
Emission Class: near IR
Detection Method: Fluorescent
Excitation Class: near IR
For Use With (Equipment): Fluorescence Microscope, Confocal Microscope, Flow Cytometer
Sub-Cellular Localization: Nucleus
Flow Cytometer Laser Lines: 633
Excitation⁄Emission (nm): 640⁄663
Shipping Condition: Room Temperature

Contents & storage

Store at ≤-20°C, desiccate and protect from light

Documents

Manuals & protocols

Recommended products