Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

Qtracker® 705 Vascular Labels

Catalog number: Q21061MP

Invitrogen™  Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Qtracker® non-targeted quantum dots are designed to be injected into the tail vein of mice for the study of vascular structure using small animal in vivo imaging (SAIVI) techniques. These nanocrystals exhibit intense fluorescence with red-shifted emission for increased tissue penetration, and have a PEG surface coating specially developed to minimize nonspecific interactions and reduce immune response by the tissue. Because the PEG surface coating does not contain reactive functional groups, the Qtracker® non-targeted quantum dots are retained in circulation longer and can be imaged for up to 3 hours with a single injection or for longer periods of time with additional injections.

Need a different emission spectrum or longer tracking? View our other mammalian cell tracking products.

Key Attributes:

Qtracker® 705 label has Ex/Em (405-665/705) nm
Designed for small animal in vivo imaging
Introduced via tail vein injection, can be imaged for up to 3 hours after injection
Available in four colors—565 nm, 655 nm, 705 nm, or 800 nm emission

Read more about SAIVI and about applications for Qdot® nanocrystals.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: Vascular Imaging Reagents
Concentration: 2 µM
Product Line: QTRACKER®, Qdot®
Product Size: 200 µL
Technique: In Vivo Imaging
Shipping Condition: Room Temperature

Contents & storage

Contains 200 µl Qtracker® non-targeted quantum dots (2 µM solution in 50 mM borate buffer, pH 8.3). Store at 2–6°C. Do not freeze. Stable for at least 6 months.

Documents

Manuals & protocols

Recommended products