BioProbes® Journal, ProbesOnline™, and BioPath™ E-newsletters. Subscribe today!

Qdot™ 705 ITK™ Carboxyl Quantum Dots

Catalog number: Q21361MP

Invitrogen™  Related applications: Cellular Imaging | Labeling Chemistry | Protein Labeling & Crosslinking | Transfection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Qdot® 705 ITK™ carboxyl quantum dots are the ideal starting material for preparing custom conjugates that require high loading of biomolecules. These materials are carboxylate functionalized and can be coupled to amine groups of proteins and modified oligonucleotides using EDC-mediated condensation. The coatings of these probes provides more binding sites than our Qdot® ITK™ amino quantum dots, but lacks PEG linkers that help to prevent non-specific interactions. These materials can be conjugated to X-PEG-amine bi-functional linkers for custom reactivity and higher specificity. Our Qdot® ITK™ carboxyl quantum dots are provided as 8 µM solutions and are available in all 9 Qdot® probe colors.

Important Features of Qdot® ITK™ Carboxyl Quantum Dots:
• Qdot® 705 ITK™ carboxyl quantum dot has emission maxima of ~705 nm
• Extremely photostable and bright fluorescence
• Efficiently excited with single-line excitation sources
• Narrow emission, large Stokes shift
• Available in multiple colors
• Ideal labeling and tracking applications


Properties of Qdot® Nanocrystals
Qdot® probes are ideal for imaging and labeling applications that require bright fluorescent signals and/or real-time tracking. Unique among fluorescent reagents, all nine available colors of Qdot® probes can be simultaneously excited with a single (UV to blue-green) light source. This property makes these reagents excellent for economical and user-friendly multiplexing applications. Qdot® labels are based on semiconductor nanotechnology and are similar in scale to moderately sized proteins.

About the Innovator’s Tool Kit Qdot® ITK™ Reagents
These Qdot® ITK™ probes are ideal for researchers who wish to prepare specific (non-stocked) conjugates for their applications and need customizable conjugation functionality.

Other Forms of Qdot® Nanocrystals are Available
In addition to the carboxyl-derivatized form, we offer Qdot® ITK™ quantum dots with amino and aliphatic hydrocarbon modifications. We’ve also developed a wide range of Qdot® nanocrystals conjugates and labeling kits. Investigate the properties of Qdot® nanocrystals or read the Molecular Probes® Handbook Section 6.6—Qdot® Nanocrystals to find out more.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Qdot® 705
Chemical Reactivity: Amine
Product Line: ITK™, Qdot®
Product Size: 250 µL
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products