Related applications: Cellular Imaging | Labeling Chemistry
Error loading your content!
Catalog number |
Select a plan
Unit size
|
Price ({{currency}}) | Availability | Qty | |
---|---|---|---|---|---|
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} |
|
— |
Label or Dye: | Qdot® 605 |
---|---|
Product Line: | ITK™, Qdot® |
Product Size: | 4 mL |
Shipping Condition: | Room Temperature |