Qubit® dsDNA HS Assay Kit

Catalog number: Q32851

Invitrogen™  Related applications: Nucleic Acid Quantitation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Qubit® dsDNA HS (High Sensitivity) Assay Kit is designed specifically for use with the Qubit® Fluorometer. The assay is highly selective for double-stranded DNA (dsDNA) over RNA and is designed to be accurate for initial sample concentrations from 10 pg/µL to 100 ng/µL. The kit provides concentrated assay reagent, dilution buffer, and pre-diluted DNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 µl and 20 µl is acceptable), and read the concentration using the Qubit® Fluorometer. Common contaminants, such as salts, free nucleotides, solvents, detergents, or protein are well tolerated in the assay.

Notes:
1. All Qubit® assay kits can be used with the Qubit® 1.0, Qubit® 2.0 and Qubit® 3.0 Fluorometers.
2. 500 µL thin-walled PCR tubes are required but not included.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type (General): dsDNA
For Use With (Equipment): Qubit® Fluorometer
Quantitation Range: 0.2-100 ng
Number of Reactions: 100 reactions
Product Line: Quant-iT™, Qubit™
Product Size: 100 assays
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Note: 500 µL thin-walled PCR tubes are required but not included.

Documents

Manuals & protocols

Recommended products