Invitrogen™

imMedia™ Growth Medium, agar, ampicillin

Catalog number:  Q60120

 Related applications: Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

imMedia Growth Medium is pre-mixed, pre-sterilized E. coli growth medium that can be prepared in just five minutes without autoclaving. imMedia Growth Medium contains everything you need to prepare low-salt LB agar plates. You'll save hours of medium preparation time because there is:

• No weighing or mixing of media components
• No autoclaving
• No waiting for media to cool before adding antibiotics

Fast medium preparation
imMedia Growth Medium is fast food for E. coli. Simply mix with water in a clean flask, heat in a microwave oven, and in less than 5 minutes your medium is prepared.

The ultimate convenience
For your convenience, imMedia Growth Medium is available for preparing liquid medium or agar plates, with or without X-gal/IPTG. You have a choice of two antibiotics: ampicillin and kanamycin. imMedia Growth Medium is packaged in ready-to-use individual pouches. Each pouch contains sufficient reagents for preparing 200 mL of liquid media or 8-10 standard 100 mm2 agar plates.

Quality results
Each type of imMedia Growth Medium is extensively tested to ensure E. coli growth, antibiotic activity, and sterility. imMedia "Blue" products (with X-gal/IPTG) are tested for efficient blue/white colony screening.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Final Product: Agar Plates
Preparation Method: Microwave
Form: Powder
Included Antibiotics: Ampicillin
Target Organism Class: E. coli
Other Additives: Heat Stabilizers
Product Line: imMedia™
Product Size: 20 each

Contents & storage

Each type of imMedia™ growth medium is provided in a 20-pouch box. Each pouch contains sufficient reagents to prepare 8-10 standard 100 mm2 agar plates.

Store at room temperature.

All reagents are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products