Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Selectively isolate and presumptively identify Pseudomonas aeruginosa and other gram-negative, non-fermentative bacteria with Thermo Scientific™ Remel™ Cetrimide Selective Agar. The composition of Cetrimide Selective Agar promotes the selective growth of P. aeruginosa1,2 and enhances the Pseudomonas-specific pigment, pyocyanin3.
Certifications/Compliance | Industrial Reference: BAM, USP. |
Description | Cetrimide Selective Agar |
Format | 12mm x 85mm Monoplate |
Product Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | dimFormat | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R01292 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Cetrimide Selective Agar is a solid medium recommended for use in qualitative procedures for selective isolation and presumptive identification of Pseudomonas aeruginosa.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
1. Lowbury, E.J. 1951. J. Clin. Pathol. 4:66–72.
2. Collins, A.G and Lowbury, E.J. 1955. J. Clin. Pathol 8:47–47.
3. King, E.O. Ward, M.K. and Raney, D.E. 1954. J. Lab. Clin. Med. 44:301–307.
4. United States Pharmacopeial Conv. Inc. 2008. United States Pharmacop. 31/The Natl. Formul. 26, Supp. 1, 8-1-08, Online. United States Pharmacopeial Conv. Rockville, Md.
5. European Directorate for the Quality of Medicines and Healthcare. 2008. The European pharmacopoeia, 6th ed., Supp. 1, 4-1-2008, online. European Directorate for the Quality of Medicines and Healthcare, Council of Europe, 226 Avenue de Colmar BP907-, F-67029 Strasbourg Cedex 1, France.