Use ProLong® Live Antifade Reagent for brightest fluorescent images with live cells

Rhod-3 Calcium Imaging Kit

Catalog number: R10145

Molecular Probes™  Related applications: Cell Structure | Cell Viability, Proliferation & Function | Cellular Imaging | Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The novel Rhod-3 Calcium Imaging Kit is designed for live-cell imaging of cytosolic calcium signaling in combination with green-fluorescent dyes or proteins.The red-fluorescent Rhod-3 AM dye supplied in the Rhod-3 Calcium Imaging Kit offers significant improvements over existing Ca2+-sensing dyes. The uptake into organelles has been reduced, which translates into better cytosolic localization. Also, Rhod-3 AM exhibits a large increase in fluorescence emission upon binding to calcium, making it more sensitive than other red-fluorescent calcium dyes.

• Compared to existing red calcium dyes, Rhod-3 AM exhibits a large increase (>2.5 fold) in fluorescence upon binding Ca2+ and very low fluorescence at rest (without Ca2+ binding).

• Narrow emission enables simultaneous detection with GFP or other dyes.

• Enhanced cytosolic localization for better detection of cytosolic transients

For easy cell loading, the Rhod-3 Calcium Imaging Kit contains PowerLoad™ Concentrate. Due to the unique nature of the PowerLoad™ solution, it can be used in the presence of complete culture media, thus reducing the negative effects of replacing media or loading in serum-free media.

A proprietary, water-soluble form of probenecid (which is commonly used to inhibit cellular transport and thus reduces the baseline signal) is also supplied with the Rhod-3 Calcium Imaging Kit.

Calcium indicators are important tools in signal transduction and cell-based pharmacological screening. In cells and tissues with blue or green autofluorescence, long-wavelength (i.e., red-shifted) calcium indicators provide a means to bypass this overlapping autofluorescence. Additionally, red-shifted calcium dyes are well suited for calcium imaging experiments multiplexed with green-fluorescent protein (GFP) or other green fluorescent dyes.

The Rhod-3 AM dye (included in the Rhod-3 Calcium Imaging Kit) is an improved red-shifted rhodamine-based calcium dye with an attached AM ester group. Once inside the cell, intracellular esterases cleave the AM group, trapping the dye in the cell and rendering the dye fluorescent. As calcium binds, the fluorescence intensity of the dye is enhanced. The spectral properties of Rhod-3 AM (ex/em = ~560/600 nm) and strong fluorescence upon calcium binding provide an excellent signal window and make it our brightest red calcium dye. Additionally, Rhod-3 AM displays a more uniform cytosolic distribution and improved signal compared to existing red calcium dyes such as Rhod-2.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Indicator Type: Calcium Indicator
Indicator Properties: Fluorescent Dye-Based
For Use With (Equipment): Fluorescence Microscope
Cell Permeability: Cell-Permeant
Form: Lyophilized
Color: Red
Product Size: 1 kit
Emission Class: Visible
Detection Method: Fluorescent
Excitation Class: Visible
Sub-Cellular Localization: Cytoplasm
Excitation⁄Emission (nm): 550⁄600
Shipping Condition: Room Temperature

Contents & storage

The kit contains sufficient reagents for staining 50 coverslips based upon the protocol provided. Store kit at -20°C, desiccated and protected from light. When stored as directed, this kit is stable for 1 year.

Documents

Manuals & protocols

Recommended products