Zeocin™ Selection Reagent

Catalog number: R25001

Gibco™  Related applications: Insect Cell Culture | Mammalian Cell Culture | Transfection | Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Gibco® Zeocin™ Selection Antibiotic is a formulation of phleomycin D1, a glycopeptide antibiotic produced by Streptomyces verticillus. Zeocin™ Selection Antibiotic acts by binding to and cleaving DNA, leading to cell death.

Zeocin™ Selection Antibiotic is used for selection of of mammalian, insect, yeast, bacterial, and plant cell cultures. It is used at a working concentration ranging between 50–400 µg/mL depending on the cell type. This product is supplied at 100 mg/mL in sterile water in 8 x 1.25 mL vials.

Other Choices and More Information
We offer a wide range of antibiotics and antimycotics in both powder and liquid formats.

See the complete list, or find products for:
Contamination control
Eukaryotic and bacterial selection

See recommendations for working concentrations for selection antibiotics.

Learn more about the use of antibiotics and antimycotics in cell culture, and review guidelines for decontaminating cultures.

Zeocin is a trademark of Invivogen.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Validated Application: Bacterial Selection, Eukaryotic Selection⁄Stable Cell Line Generation
Agent: Zeocin™
Form: Liquid
Reagent Type: Antibiotic (Selective)
Concentration: 100 mg⁄ml
Product Line: Zeocin™
Product Size: 8 x 1.25 mL

Contents & storage

Storage conditions: -10 to -30°C
Shipping conditions: Ice
Shelf life: 24 months from date of manufacture

Documents

Manuals & protocols

Recommended products