Get the sharpest, brightest images. Try new Invitrogen™ ProLong™ Glass antifade mountant with glass-matching refractive index of 1.52. Learn more
Invitrogen™

Image-iT™ Fixation/Permeabilization Kit

Catalog number:  R37602

 Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Image-iT® Fixation/Permeabilization Kit is a convenient-to-use fixation/permeabilization kit that contains all the necessary reagents to prepare your cells for antibody staining and imaging. The high quality components help preserve cell morphology and reduce background staining and come in a practical box with single-use vials and easy-to-follow protocols. Note: the 4% formadehyde, methanol-free fixative solution in this kit is also available for separate purchase (see Cat. No. R37814).

• A complete fixation/permeabilization kit with ready-to-use components and room temperature storage
• Optimized for secondary antibody labeling, fixed-cell dye staining, and GFP applications
• Reduced background staining

See other ReadyProbes® reagents for cell staining
Find more tools for fixed cell imaging

The Image-iT® Fixation/Permeabilization Kit Contains:
Fixative: High purity 4% formaldehyde in PBS, pH=7.3
Permeabilization solution: 0.5% Triton X-100
Blocking buffer: 3% BSA, fraction V, de-lipidated, New Zealand source, in DPBS
Wash solution: PBS, pH 7.4

Figures

Specifications

Product Line: Image-iT®, Molecular Probes®
Product Size: 1 kit
Reagent Type: Fixation Reagent, Permeabilization Reagent, Triton X-100
Shipping Condition: Room Temperature

Contents & storage

Fixative Solution: 20 mL
Permeabilization solution: 20 mL
Blocking solution: 12 x 10 mL
Wash Buffer: 500 mL

Store at ≤25 °C.

Documents

Manuals & protocols

Recommended products