Invitrogen™

Rhodamine 110, bis-(CBZ-L-Isoleucyl-L-Prolyl-L-Arginine Amide), Dihydrochloride (BZiPAR)

Catalog number:  R6505

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Molecular Probes® bisamide derivative of rhodamine 110 (R110) is a sensitive and selective substrate for assaying proteases in solution or inside living cells. These fluorogenic substrates contain an amino acid or peptide covalently linked to each of R110’s amino groups. Upon enzymatic cleavage, the nonfluorescent bisamide substrate is converted first to the fluorescent monoamide and then to R110, with a further increase in fluorescence. The substrate can be used to continuously measure enzyme activity in cell extracts and purified enzyme preparations using a fluorometer or fluorescence microplate reader, or can be used for intracellular protease assays with analysis by flow cytometry or fluorescence microscopy.

Substrate Specifications:
• Substrate name: (CBZ-Ile-Pro-Arg)2-R110
• Useful to measure: Trypsin
• Ex/Em of R110 product: (498/521 nm)

Properties of the R110 Reaction Product
The R110 cleavage product has spectral properties similar to those of fluorescein, making it compatible with standard fluorescein filter sets. Because of the large extinction coefficient of R110, signal to background ratios are very good. And, unlike fluorescein, the fluorescence intensity of the rhodamine 110 reaction product is constant from pH 3–9. We also offer an R110 reference standard for use in your assays.

We’ll Make a Custom R110 Substrate for You
If you can’t find what you’re looking for in our stocked list, we’ll prepare a custom R110 substrate for you. Our custom services are efficient and confidential, and we stand by the quality of our work. We are ISO 9001:2000 certified.

For more information about these products, read Detecting Peptidases and Proteases—Section 10.4 in the Molecular Probes® Handbook.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Permeability: Cell-Permeant
Detection Method: Fluorescent
For Use With (Equipment): Flow Cytometer, Fluorescence Microscope
Product Size: 5 mg
Substrate: BZiPAR
Substrate Properties: Peptide-Based Substrate
Substrate Type: Caspase Substrate
Target Enzyme: Trypsin
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols