T-REx™-293 Cell Line

Catalog number: R71007

Invitrogen™  Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

T-REx™ Cell Lines stably express the tetracycline repressor protein (Table 1). They save significant time and effort when using the T-REx™ System. The T-REx™ Cell Lines are functionally tested by transient transfection with the positive control vector pcDNA™4⁄TO⁄lacZ. T-REx™ Cell Lines exhibit extremely low basal expression levels in the repressed state and high expression upon induction with tetracycline (Figure 1).
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Species: Human
Cell Line: T-REx™-293 Cell Line
System: T-Rex™
Product Size: 3 x 106 cells

Contents & storage

3 x 106 cells are supplied frozen in 1 ml 90% complete medium and 10% DMSO. Store in liquid nitrogen. Cells are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols