Flp-In™-293 Cell Line

Catalog number: R75007

Invitrogen™  Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Flp-In™ 293 cell line is designed for rapid generation of stable cell lines that ensure high level expression of your protein of interest from a Flp-In™ expression vector. These cells contain a single stably integrated FRT site at a transcriptionally active genomic locus. Targeted integration of a Flp-In™ expression vector ensures high-level expression of your gene of interest. Flp-In™-293 cell line was created by transfecting the parent cell lines with pFRT⁄lacZeo and selecting for stable Zeocin™- resistant clones.

Flp-In™-293 cells work well with Flp-In™ vectors that express a gene from the CMV promoter (e.g., pcDNA™5⁄FRT, pcDNA5⁄FRT⁄V5-His-TOPO®, and pSecTag⁄FRT⁄V5-His-TOPO
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Species: Human
Cell Line: Flp-In™-293
System: Flp-In™
Product Size: 1 mL

Contents & storage

1 x 10E7 cells are supplied frozen in 1 ml of 90% complete medium and 10% DMSO. Cells must be stored in liquid nitrogen. Cells are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products