BG01V/hOG Human Embryonic Stem Cells (hESCs)

Catalog number: R7799105

Gibco™  Related applications: Mammalian Cell Culture | Stem Cell Culture | Stem Cell Engineering & Reprogramming

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Variant hESC hOct4-GFP Reporter Cells

BG01V/hOG Cells are derived from the BG01V human embryonic stem cell (hESC) line (ATCC No. SCRC-2002) (Mitalipova et al. (2003); Plaia et al. (2006)). BG01V cells stain positively for pluripotency markers and can differentiate to representatives of all three primary germ layers. An integration vector containing an EmGFP (Emerald Green Fluorescent Protein) gene directed by the human Oct4 promoter and followed by the HSV-TK polyadenylation signal was stably integrated into the genome of BG01V cells. The engineered cells express EmGFP under the appropriate conditions when the Oct4 promoter is active. This promoter is only active when hESCs are in the pluripotent state and therefore acts as a sensitive indicator of differentiation (Pan et al. (2002)).

Applications:
1. BG01V/hOG Cells are engineered to enable monitoring of pluripotency of hESCs without sacrificing cells. When pluripotent, these cells express EmGFP. EmGFP expression is lost upon differentiation.
2. BG01V/hOG Cells can be used as a reporter of the culture’s response to external stimuli including media composition, stress, and whether or not culture conditions are adequate to maintain hESCs in an undifferentiated state.
3. BG01V/hOG Cells can be used to determine the efficiency of your differentiation assays.

Intended use:
Research use only. Caution: Not intended for human or animal diagnostic or therapeutic uses.

Contents and Storage:
The vial of BG01V/hOG Cells should be stored in liquid nitrogen.

References:
1. Mitalipova, M. et al. (2003) Stem Cells 21: 521-526
2. Pan, G. J. et al. (2002) Cell Res 12: 321-329
3. Plaia, T. W. et al. (2006) Stem Cells 24: 531-546

Useful links:
User Manual for BG01V/hOG Cells
Embryonic Stem Cell (ESC) Product Overview Sheet
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Form: Cryopreserved
Species: Human
Cell Line: BG01V/hOG
Cell Type: Stem Cells (Embryonic)
Product Size: 2 x 106 cells
Parental Cells: BG01V hESC Line
Stably Integrated Gene or Element: Human Oct4 Promoter-EmGFP

Documents

Manuals & protocols

Recommended products