ProBond™ Nickel-Chelating Resin

Catalog number: R80101

Novex™  Related applications: Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

ProBond™ Nickel-Chelating Resin is a nickel-charged affinity resin used to purify recombinant proteins containing a polyhistidine (6xHis) sequence. Proteins bound to the resin may be eluted with either low pH buffer or by competition with imidazole or histidine. One-step purification can be performed under both native and denaturing conditions. ProBond™ Resin uses the chelating ligand iminodiacetic acid (IDA) coupled to a highly cross-linked 6% agarose resin that is suitable for use in FPLC, batch, and gravity flow applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: Suspension
Column Type: Affinity Column
Product Size: 50 mL
Immobilized Phase: Nickel-Chelating

Contents & storage

The ProBond™ resin is pre-charged and able to bind 1-5 mg of recombinant protein per 1 ml of resin. It is provided as a 50% slurry in 20% ethanol. The resin will appear blue in color when charged with Ni2+ . Store at +4°C. ProBond™ resin is guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products