CD62L / L-Selectin Antibody, Pacific Blue™ conjugate (MEL-14)

Catalog number: RM4328

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CD62L / L-Selectin Monoclonal Antibody for Flow
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Mouse
Validated Application: Flow Cytometry
Host Species: Rat
SubClass (Isotype): IgG2a, IgG2b, IgG2c
Clonality: Monoclonal
Clone ID: MEL-14
Antibody Fragment: Whole Antibody
Target Gene: CD62L
Target Molecule: Protein
Gene Symbol: SELL
Label or Dye: Pacific Blue™
Excitation⁄Emission (nm): 405⁄455
Purification: Affinity Purified
Concentration: 100 µg⁄ml
Form: Liquid
For Use With (Equipment): Flow Cytometer
Product Size: 1 mL
Quantity: 1 each
Shipping Condition: Wet Ice

Contents & storage

CD62L, Rat anti- mouse, (Pacific Blue™), 1 mL in PBS with 0.1% sodium azide. Store reagents at 2-8°C.

Documents

Manuals & protocols

Recommended products