BioProbes® Journal, ProbesOnline™, and BioPath™ E-newsletters. Subscribe today!

Streptavidin, Marina Blue® conjugate

Catalog number: S11221

Molecular Probes™

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Our streptavidin, Marina Blue™ conjugate is a bright, blue-fluorescent probe with excitation/emission ~365/460 nm. Streptavidin is a nonglycosylated biotin-binding protein that is isolated from bacteria and has a near-neutral isoelectric point. It is widely used to detect biotinylated probes, because it reportedly exhibits less nonspecific binding than does the glycosylated, biotin-binding protein, avidin.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Marina Blue®
Product Size: 1 mg
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products