Invitrogen™

SYTO™ Orange Fluorescent Nucleic Acid Stain Sampler Kit - SYTO™ dyes 80-85 - 50 μL each

Catalog number: S11360
Invitrogen™

SYTO™ Orange Fluorescent Nucleic Acid Stain Sampler Kit - SYTO™ dyes 80-85 - 50 μL each

Catalog number: S11360
Catalog Number
S11360
Unit Size
1 kit
Price (USD)
Price: 470.00
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S113601 kit
Price: 470.00
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The SYTO Orange Fluorescent Nucleic Acid Stain Sampler Kit contains our collection of cell-permeant, orange-fluorescent SYTO nucleic acid stains. Because the dyes may demonstrate different staining behaviors with various tissues and cells, it may be necessary to test the dyes to find the optimal dye for a specific application. The kit contains 50 μL each of SYTO 80–85 dyes.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Orange
Excitation Wavelength Range
531, 530, 541, 543, 567, 567 nm
Dye Type
Cell-Permeant
For Use With (Equipment)
Fluorescence Microscope
Volume (Metric)
50 μL
Detection Method
Fluorescence
Description
SYTO™ Orange Fluorescent Nucleic Acid Stain Sampler Kit - SYTO™ dyes 80-85, 50 μL each
Emission
545, 544, 560, 559, 582, 583 nm
Product Line
SYTO
Shipping Condition
Room Temperature
Label Type
Fluorescent Dye
Product Type
Stains Dimer Sampler Kit
SubCellular Localization
Nucleic Acids

Contents & Storage

Store in freezer at -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text