Streptavidin, Allophycocyanin Conjugate, premium grade

Catalog number: S32362

Molecular Probes™

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Streptavidin is a bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than does avidin. Allophycocyanin is an intensely bright phycobiliprotein with a broad absorption profile that peaks at ~650 nm, and a fluorescence emission maximum of ~660 nm. Our streptavidin allophycocyanin conjugate *premium grade* is a further fractionation of our S-868 product, intended especially for applications that benefit from additional purity.

View a selection guide for all streptavidin RPE, APC, and tandem conjugates.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Fluorescent
Flow Cytometer Laser Lines: 633 or 635
Label or Dye: APC (Allophycocyanin)
Form: Solution
Product Size: 250 µL
Concentration: 1 mg⁄ml
Shipping Condition: Wet Ice
For Use With (Equipment): Flow Cytometry, Immunoassays, Blot Analysis, Histochemical Applications
Excitation⁄Emission (nm): 650⁄660

Contents & storage

Contains 1 vial of streptavidin APC, premium grade, conjugate (1 mg/mL).

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products