Invitrogen™

Safe Imager™ Viewing Glasses

Catalog number:  S37103

 Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Safe Imager™ viewing glasses are for use with the Safe Imager™ blue-light transilluminator. These glasses allow the user to visualize gel bands without the use of the Safe Imager™ amber filter, which facilitates band excision.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Safe Imager™
Product Line: SYBR Safe™
Product Size: 1 each
Type: Tools
Shipping Condition: Room Temperature

Contents & storage

I pair of Safe Imager™ viewing glasses

Documents

Manuals & protocols

Recommended products