Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

Streptavidin-Horseradish Peroxidase (HRP) Conjugate

Catalog number: SA10001

Molecular Probes™

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Streptavidin-HRP conjugate is typically used in the immunodetection of biotinylated proteins. This conjugate is suitable for use in immunoblotting, ELISA, immunomicroscopy. The amount of conjugate provided is sufficient for 25 western blots using 10 ml of working solution per reaction or 25 immunostaining reactions using 1 ml of working solution per reaction as appropriate.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 50 µL

Contents & storage

Store in refrigerator (2—8°C).

Documents

Manuals & protocols

Recommended products