BioProbes® Journal, ProbesOnline™, and BioPath™ E-newsletters. Subscribe today!

Streptavidin-Fluorescein Isothiocyanate (FITC) Conjugate

Catalog number: SA10002

Molecular Probes™

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Streptavidin-FITC conjugate is typically used in the immunodetection of biotinylated proteins. This conjugate is suitable for use in immunomicroscopy and fluorescence-activated cell sorting and analysis. The amount of conjugate provided is sufficient for 25 immunostaining reactions using 1 ml of working solution per reaction as appropriate.
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Label or Dye: FITC (Fluorescein)
Product Size: 50 µL

Contents & storage

Store in refrigerator (2—8°C).

Documents

Manuals & protocols

Recommended products