Novex™ Semi-Dry Blotter

Numéro de catalogue:  SD1000

Invitrogen™  Applications liées: Protein Assays and Analysis | Protein Gel Electrophoresis

Error lors du chargement de votre contenu!

  Numéro de catalogue
Choisir votre mod�le de prix
Taille unitaire
Prix ({{currency}}) Disponibilité Qté
{{product.sku}} {{product.sku}} également connu comme {{product.formattedSku}} 
{{product.availability.message}}
Ajouts

Votre stock sur site

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} En stock)
›› {{supplyCenter.scName}}(En rupture de stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Haut de la page

Description

The Novex® Semi-Dry Blotter quickly and efficiently transfers proteins and nucleic acids from mini and midi gels to membranes. Simultaneous transfer can be achieved from up to 2 midi gels, 4 mini gels, or 2 E-PAGE™ gels. The Novex® Semi-Dry Blotter is designed for fast set-up and ease of use. The platinum-coated titanium anode plate and stainless steel cathode plate provide fast and efficient electroblotting using less transfer buffer than wet (tank) blotting methods.

The Novex® Semi-Dry Blotter is designed for convenience and efficiency:
• Economical – Horizontal configuration requires less than 200 ml of buffer to transfer
• High-capacity – A large, 20 x 20 cm transfer surface allows up to two midi gels or four mini gels to be transferred at one time
• Flexible – Field strength across the blot is maximized and evenly distributed, for efficient and complete transfers regardless of the number and size of gels blotted
• Durable – Stainless steel and platinum-coated titanium electrodes allow for corrosion-free operation
• Fast – Blotting is typically completed in 30-60 min

A Complete Western Blotting System
We offer precut (8.3 cm × 7.3 cm) and pre-assembled membranes and filter paper sandwiches (available separately) to further simplify and speed the western blotting process. Choose between Invitrolon™ PVDF (LC2002 or LC2005), or nitrocellulose (LC2000 or LC2001) containing sandwiches.

The Novex® Semi-Dry Blotter requires an external power supply for operation (up to 800 mA, depending on the number and size of blots).
For Research Use Only. Not for use in diagnostic procedures.

Spécifications

Mode of Transfer: Semi-Dry
Gel Compatibility: Novex® Midi Gels, Novex® Mini Gels, NuPAGE® Gels
Running Dimension: Horizontal
For Use With (Equipment): Novex® Semi-Dry Blotter
Capacity: Up to 2 E-PAGE gels, Up to 2 midi-gels, Up to 4 mini-gels
Gel Size: Midi (8 cm x 13 cm), Mini (8 cm x 8 cm)
Product Line: Novex™
Product Size: 1 each
Shipping Condition: Room Temperature

Contenu et stockage

Store at room temperature.

Documents

Manuels et protocoles