BioProbes® Journal, ProbesOnline™, and BioPath™ E-newsletters. Subscribe today!

Streptavidin FITC

Catalog number: SNN1008

Molecular Probes™

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Purified from Streptomyces avidinii and provided in a phosphate buffered saline solution. The streptavidin/avidin-biotin staining and detection strategy is used when increased assay sensitivity is desired for a wide variety of systems.

Format/Formulation: FITC Conjugate
For Laboratory Use

Specifications

Label or Dye: FITC (Fluorescein)
Product Size: 1 mg
Shipping Condition: Wet Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products