Galacto-Star™ β-Galactosidase Reporter Gene Assay System for Yeast Cells

Catalog number: T1019

Novex™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Galacto-Star™ One-Step beta-Galactosidase Reporter Gene Assay System chemiluminescent reporter gene assay system enables rapid, sensitive detection of β-galactosidase in yeast cell lysates.

• Glow assay kinetics provides a "window" during which measurements may be performed, facilitating HTS applications where assay automation is used.
• Can be used with either mammalian or yeast model systems, providing flexibility in choice of model systems.
• Wide dynamic range of β-galactosidase assay lets the user measure enzyme level accurately from femtogram to nanogram range.
• Assay sensitivity is 100- to 1,000-fold better than either the isotopic⁄non-isotopic assays for chloramphenicol acetyl transferase (CAT) or the colorimetric⁄fluorescent assays for β-galactosidase, providing greater sensitivity than competing assays.
• Highly sensitive assay with a wide dynamic range permits detection of high and low levels of reporter without performing numerous sample dilutions.
• Non-radioactive reporter gene assay kit eliminates concerns over use of radioisotopes
• Assay can be completed in about one hour, providing fast assay turnaround.

Sensitive
The chemiluminescent assay exhibits over three orders of magnitude greater sensitivity than colorimetric β-galactosidase assays and can be completed in a fraction of the time required for assays for chloramphenicol acetyltransferase (CAT) and ELISA-based reporter assays.

Versatile
The Galacto-Star™ system includes Galacton-Star® chemiluminescent substrate for β-galactosidase and is suited for measurement in both luminometers and scintillation counters. Automatic injection is not required.

Long-Lasting Signal
Light emission from the Galacto-Star™ assay reaches maximum in 60-90 minutes and remains constant for nearly one hour. Cell lysate is mixed with reaction buffer containing Galacton-Star® substrate and Sapphire-II™ luminescence enhancer. After a 60-90 minute incubation at room temperature, the signal is measured in a luminometer or scintillation counter.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Format: 96-well plate
Readout: End Point
Substrate: Galacton-Star®
Technique: Enhanced Chemiluminescence
Assay Type: β-Gal (lacZ) Assay
Product Size: 600 assays
Target Enzyme: Beta-Galactosidase
Substrate Type: Beta-Gal Substrate
Compatible Cells: Yeast Cells
Detection Method: Chemiluminescent
Substrate Properties: Chemical Substrate
Sample Type (Specific): Cell Cultures
For Use With (Equipment): Luminometer (Microplate)
High Throughput Compatibility: High Throughput-Compatible

Contents & storage

This kit contains the lysis solution, substrate, enhancer, and required buffer. Store the product at 4°C.

Documents

Manuals & protocols