Invitrogen™

Western-Star™ Immunodetection System with Goat Anti-Mouse IgG + IgM AP conjugate

Catalog number:  T1046

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Western-SuperStar™ Immunodetection System is a high sensitivity, high signal intensity,
chemiluminescence-based immunodetection system that provides speed, flexibility, control of film exposure time, and the ability to use multiple imaging methods.

• Most complete western blot detection kit available
• Image immediately or up to 24 hours later
• Compatible with multiple membrane types, including PVDF, nitrocellulose, and nylon
• High signal intensity – ideal for X-ray film and CCD-based imaging
• High sensitivity and high signal to noise – detect femtogram levels of protein
• Strip, reprobe, and image at least three times – detect multiple proteins
• Perform phosphoepitope detection

The Western-SuperStar™ System is ideal for confirmation of protein knockdown in siRNA-mediated gene silencing experiments.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Chemiluminescent
For Use With (Equipment): Phosphorimager, Photographic Imager, X-Ray Film
Membrane Compatibility: Nitrocellulose, Nylon, PVDF
Product Line: Western-Star™
Product Size: 30 blots
Reactivity: Mouse
Sample Type (Specific): Protein
Substrate: CDP-Star®
Substrate Type: AP (Alkaline Phosphatase) Substrate
Target Isotype: IgG, IgM

Contents & storage

Includes Western-SuperStar™ Reagent Set (containing Blocker, Wash Buffer, Assay Buffer, Substrate, Enhancer, Blot-Loc™ bags, Development Folders and pair of Tweezers) and an anti-rabbit secondary antibody conjugated to alkaline phosphatase. Store all liquid reagents at 2-8° C.

Documents

Manuals & protocols