NEW - Major Software Update Available! — Cell Cycle Analysis can be performed using the Tali® Image-Based Cytometer

Tali™ Image-Based Cytometer Power Cord

Catalog number: T10793

Invitrogen™  Related applications: Cell Counting, Viability, & Cryopreservation | Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This power cord is a replacement part for the Tali® Image-Based Cytometer instrument. The Tali® Image-Based Cytometer Power cord and the four universal adaptors included have been specifically designed for use with the Tali® Image-Based Cytometer instrument. This product has been CE certified.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Tali® Image-Based Cytometer
Product Line: Tali®
Product Size: 1 each
Shipping Condition: Room Temperature
Type: Power Cord

Contents & storage

1 replacement power cord for the Tali® Image Based Cytometer.

Includes 4 adapter plugs (for U.S./E.U./U.K./Australia).

Documents

Manuals & protocols