NEW - Major Software Update Available!—Cell Cycle Analysis can be performed using the Tali® Image-Based Cytometer
NEW - Tali® Cell Cycle Kit Launched! Read More!

Tali® Cellular Analysis Slides

Catalog number: T10794

Invitrogen™  Related applications: Cell Counting, Viability, & Cryopreservation | Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Tali® Cellular Analysis Slides are specifically designed for use with the Tali® Image-Based Cytometer. Each slide contains two separate, enclosed analysis chambers designed to accommodate 25 µl of sample per chamber. Each box contains 50 slides.

Ready to Use for Your Convenience
Each Tali® Cellular Analysis slide is pre-sterilized and individually packaged. Since the Tali® Image-Based Cytometer is a fluorescent cellular analysis instrument the slides are composed of low fluorescent plastic. The slides are disposable; after use the slides can be disposed of with you regular biohazard waste.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Learn more about the the Tali® Image-based Cytometer
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Tali® Image-Based Cytometer
Product Line: Tali®
Product Size: 50 slides
Shipping Condition: Room Temperature
Type: Slides

Contents & storage

Contains one box of 50 slides. Store at room temperature.

Documents

Manuals & protocols