Get the sharpest, brightest images. Try new Invitrogen™ ProLong™ Glass antifade mountant with glass-matching refractive index of 1.52. Learn more
Invitrogen™

TSA™ Kit #6, with HRP—Goat Anti-Mouse IgG and Alexa Fluor™ 647 Tyramide

Catalog number:  T20916

 Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

New SuperBoost™ Tyramide signal Amplification Kits offer 2-10 times the sensitivity of TSA™ kits. Explore the advantages of Alexa Fluor™ 647 Tyramide SuperBoost™ Kit - Goat anti-Mouse IgG, which is replacing TSA™ Kit #6, with HRP--Goat Anti-Mouse IgG and Alexa Fluor® 647 Tyramide.

Tyramide signal amplification is an enzyme-mediated detection method that uses the catalytic activity of horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid sequence in situ. The TSA Kit #6 includes HRP—goat anti-mouse IgG and our bright, far-red—fluorescent Alexa Fluor 647 tyramide (which is spectrally similar to Cy5 dye).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Alexa Fluor™ 647, HRP (Horseradish Peroxidase)
Product Line: Alexa Fluor™, TSA™
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products