Use Prolong Diamond with any Fluorescent Dye or Fluorescent Protein for brightest images. Learn more.

TSA™ Kit #12, with HRP—Goat Anti-Rabbit IgG and Alexa Fluor® 488 Tyramide

Catalog number: T20922

Molecular Probes™  Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Tyramide signal amplification is an enzyme-mediated detection method that uses the catalytic activity of horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid sequence in situ. The TSA Kit #12 includes HRP—goat anti—rabbit IgG and our exceptionally bright, photostable, green-fluorescent Alexa Fluor 488 tyramide (which is spectrally similar to fluorescein).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Alexa Fluor™ 488, HRP (Horseradish Peroxidase)
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products