Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits

Ulysis™ Alexa Fluor™ 647 Nucleic Acid Labeling Kit

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

ULYSIS® Nucleic Acid Labeling Kits provide a unique method to attach a fluorescent dye to nucleic acids. The labeling reagent in the kit reacts with the N7 of guanine to form a stable coordination complex, and the reaction is simple and fast—just heat denature DNA (5 minutes), add the label (react for 15 minutes), then purify.

ULYSIS® Nucleic Acid Labeling Specifications:
• Dye (Ex/Em): Alexa Fluor® 647 (650/670 nm)
• Labeling reaction is complete in as little as 15 minutes
• Available in several Alexa Fluor® dye colors


The Resulting Labeled Probe is useful for:
• Dot, northern, and Southern blots
• RNA and DNA in situ hybridization
• Multicolor fluorescence in situ hybridization (mFISH)
• Comparative genome hybridization (CGH)
• Microarray analysis

Reliable Labeling With the Universal Linkage System
We developed this series of ULYSIS® kits to enable rapid and simple coupling of our Alexa Fluor® dyes to purine bases in nucleic acid polymers. The method, the Universal Linkage System (ULS™), is based on the use of a platinum dye complex (owned by KREATECH Diagnostics) that forms a stable adduct with the N7 position of guanine and, to a lesser extent, adenine bases in DNA, RNA, PNA, and oligonucleotides. The result is a reliable nonenzymatic method for nucleic acid labeling.

Labeling is Fast and Easy
The labeling reaction typically takes only 15 minutes, and separation of the labeled nucleic acids from the unreacted ULS™ complex can be accomplished through the use of a simple spin-column procedure. DNA longer than ~1,000 base pairs requires a 10-minute DNase digestion before labeling, which both optimizes labeling and fragments the probe for efficient hybridization.

More Options for Nucleic Acid Labeling
To review various options for nucleic acid labeling, review Labeling Oligonucleotides and Nucleic Acids—Section 8.2 in the Molecular Probes® Handbook, read Nucleic Acid Amplification & Expression Profiling, or view a list of our kits.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Fluorescent
Final Product: Oligos (Labeled), Probes (Labeled DNA), Probes (Labeled RNA)
Includes Label or Dye: Yes
Label or Dye: Alexa Fluor™ 647
Labeling Method: Direct Labeling
Labeling Target: DNA (General), Oligos, RNA (General)
Product Line: Alexa Fluor™, Ulysis™
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products