Single-Channel Dead Cell Apoptosis Kit with Annexin V Alexa Fluor® 488 and SYTOX® Green Dyes, for flow cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This product detects the externalization of phosphatidylserine in apoptotic cells using recombinant annexin V conjugated to green-fluorescent Alexa Fluor™ 488 dye and dead cells using SYTOX™ Green nucleic acid stain. After treatment with both probes, apoptotic cells show green fluorescence, dead cells show brighter green fluorescence, and live cells show little or no fluorescence. These populations can easily be distinguished in the FL1 channel of a flow cytometer, freeing the other channels for detection of other fluorescent colors.

View a selection guide for all apoptosis assays for flow cytometry.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Fluorescent
Excitation⁄Emission (nm): Alexa Fluor® 488: 499⁄521, SYTOX® green: 503⁄524
For Use With (Equipment): Flow Cytometer
Format: Tube(s)
Label or Dye: Alexa Fluor™ 488, SYTOX® Green
Number of Reactions: 50
Product Line: Alexa Fluor®, SYTOX®
Product Size: 1 kit
Shipping Condition: Wet Ice

Contents & storage

Contains 1 vial of annexin V, Alexa Fluor® 488 conjugate (250 µL), 1 vial of SYTOX® green (100 µL), and 1 bottle of annexin binding buffer (5X solution, 50 mL).

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products