Invitrogen™

pPICZα A, B, & C Pichia Vectors

Catalog number:  V19520

 Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

pPICα A, B, and C vectors are 3.6 kb vectors used to express and secrete recombinant proteins in Pichia pastoris. Recombinant proteins are expressed as fusions to an N-terminal peptide encoding the Saccharomyces cerevisiae á-factor secretion signal. These vectors allow high-level, methanol inducible expression of the gene of interest in Pichia, and can be used in any Pichia strain including X-33, SMD1168H, and KM71H. pPICα vectors contain the following elements:


• Contains AOX1 promoter for tightly regulated, methanol-induced expression of the gene of interest
• All three reading frames (A, B, C versions) are provided to facilitate in-frame cloning with the C-terminal peptide
• α-factor secretion signal for directing secreted expression of the recombinant protein
• Zeocin resistance gene for selection in both E. coli and Pichia
• C-terminal peptide containing the c-myc epitope and a polyhistidine (6xHis) tag for detection and purification of a recombinant fusion protein
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Antibiotic Resistance (Bacterial): Zeocin™ (ZeoR)
Cloning Method: Restriction Enzyme ⁄ MCS
Product Size: 20 µg
Promoter: AOX1
Protein Tag or Fusion: His Tag (6x), c-Myc Epitope Tag
Vector Type: pPIC
Shipping Condition: Room Temperature

Contents & storage

20 µg each of lyophilized pPICZ A, B, and C is supplied (Store at -20C)

Documents

Manuals & protocols

Recommended products