New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.

pGAPZα A, B, & C Pichia pastoris Expression Vectors

Catalog number:  V20520

Invitrogen™  Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


pGAPZ A, B, & C and pGAPZ A, B, & C are expression vectors designed for high-level, constitutive expression in Pichia pastoris. pGAPZ and pGAPZ were created by replacing the methanol-regulated AOX1 promoter with the constitutive, glyceraldehyde-3-phosphate dehydrogenase (GAP) promoter in the backbone of the pPICZ vectors. Although the yield
of any protein constitutively expressed in the Pichia system is dependent on the toxicity of the protein to yeast, constitutive expression under the control of pGAPZ or pGAPZ vectors can sometimes produce greater yields than inducible expression (1).
For Research Use Only. Not for use in diagnostic procedures.



Antibiotic Resistance (Bacterial): Zeocin™ (ZeoR)
Cloning Method: Restriction Enzyme ⁄ MCS
Product Size: 20 µg
Promoter: GAP
Protein Tag or Fusion: His Tag (6x), c-Myc Epitope Tag
Vector Type: pGAP

Contents & storage

All vectors are supercoiled and are provided lyophilized (store at -20°C). A lacZ positive expression control strain is provided with the pGAPZ vector. Store strains at +4°C. All reagents are guaranteed stable for 6 months when properly stored.


Manuals & protocols

Recommended products