Related applications: Protein Expression
Error loading your content!
Catalog number |
Select a plan
Unit size
|
Price ({{currency}}) | Availability | Qty | |
---|---|---|---|---|---|
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} |
|
— |
Antibiotic Resistance (Bacterial): | Blasticidin (BsdR) |
---|---|
Cloning Method: | Restriction Enzyme ⁄ MCS |
Product Size: | 20 µg |
Promoter: | AOX1 |
Protein Tag or Fusion: | His Tag (6x), c-Myc Epitope Tag |
Vector Type: | pPIC |