Invitrogen™

pcDNA™6/V5-His A, B, & C Mammalian Expression Vectors

Catalog number:  V22020

 Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

All pcDNA™ vectors contain a strong promoter for high-level expression in mammalian cells, a choice of selection marker for generating stable cell lines, and an epitope tag for easy detection with a monoclonal antibody and rapid purification on nickel-chelating resin. Each vector is available in three reading frames to simplify cloning in-frame with the fusion tag.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Promoter: CMV
Key Function: Constitutive Expression
Protein Tag or Fusion: His Tag (6x), V5 Epitope Tag
Delivery Method: Transfection
Cloning Method: Restriction Enzyme ⁄ MCS
Selection Agent (Eukaryotic): Blasticidin
Vector Type: pcDNA
Constitutive or Inducible System: Constitutive
Product Line: pcDNA™
Product Size: 20 µg

Contents & storage

Vectors (and appropriate expression control) are supplied supercoiled and lyophilized. Stored at -20°C. All vectors are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols