pVAX1 Vector

Catalog number:  V26020

Invitrogen™  Related applications: Protein Expression | Viral Vaccine Production

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

pVAX1 is specifically designed for use in the development of DNA vaccines. pVAX1 has the following features:

• Eukaryotic DNA sequences limited to those required for expression in order to minimize the possibility of chromosomal integration
• Kanamycin resistance gene for selection in E. colibecause ampicillin has been reported to cause an allergic response in some individuals
• Expression levels of recombinant proteins from pVAX1 comparable to those achieved with its parent vector, pcDNA™3.1

The small size of pVAX1 and the variety of unique cloning sites simplify subcloning of even very large DNA fragments.
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Promoter: CMV
Protein Tag or Fusion: Untagged
Delivery Method: Transfection
Cloning Method: Restriction Enzyme ⁄ MCS
Selection Agent (Eukaryotic): None
Vector Type: pVAX1
Constitutive or Inducible System: Constitutive
Product Line: pVAX™
Product Size: 20 µg

Contents & storage

20 µg of each pVAX1 and expression control vector are supplied supercoiled and lyophilized. Store at -20°C. Vector is guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products