Metabolic Activity Dead Cell Apoptosis Kit with C12 Resazurin, Annexin V APC, and SYTOX™ Green, for flow cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This flow cytometry product provides a three-color fluorescence assay that distinguishes live, apoptotic, and late apoptotic cells from one another. These populations can easily be distinguished using a flow cytometer that has both 488 nm and 633 nm excitation sources (an argon-ion laser and a HeNe laser) and the following reagents Annexin V to detect phosphatidylserine, C12 resazurin for cell metabolism, and SYTOX™ Green nucleic acid stain for compromised membranes.

View a selection guide for all apoptosis assays for flow cytometry.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

LoyaltyEligible: SKUEligibleForLoyalty
Detection Method: Fluorescent
Excitation⁄Emission (nm): APC: 650⁄660, C12-Resazurin: 571⁄585, SYTOX® green: 503⁄524
For Use With (Equipment): Flow Cytometer, Fluorescence Microscope
Format: Slide(s), Tube(s)
Label or Dye: APC (Allophycocyanin), C12-resazurin, SYTOX® Green
Number of Reactions: 50
Product Line: SYTOX®
Product Size: 1 kit
Shipping Condition: Wet Ice

Contents & storage

Contains 1 vial of annexin V, APC conjugate (250 µL), 1 vial of SYTOX® green stain (100 µL), 1 vial of C12-resazurin (40 µg), and 1 vial of DMSO (1.5 mL).

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products