pRSET A, B, & C Bacterial Expression Vectors

Catalog number:  V35120

Invitrogen™  Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The pRSET vector is designed for high-level prokaryotic expression controlled by the strong bacteriophage T7 promoter. Expression is induced by the production of T7 RNA polymerase in BL21(DE3) E. coli. These cells also produce T7 lysozyme to reduce basal expression of target genes. The pRSET vector offers:

• Bacteriophage T7 promoter for high-level expression
• T7 gene 10 sequence to provide protein stability
• N-terminal polyhistidine (6xHis) tag for rapid purification with nickel-chelating resin and detection with an Anti-HisG Antibody
• N-terminal Xpress™ epitope for detection with the Anti-Xpress™ Antibody
• Enterokinase cleavage site for removal of fusion tag

A set of three vectors is provided (A, B, and C). Each has the N-terminal tag coding sequence in a different reading frame relative to the multiple cloning site to simplify in-frame cloning of your gene.
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Vector Type: pRSET
Cloning Method: Restriction Enzyme ⁄ MCS
Promoter: T7
Protein Tag or Fusion: His Tag (6x)
Antibiotic Resistance (Bacterial): Ampicillin (AmpR)
Cleavage: EK (Enterokinase) Recognition Site
Constitutive or Inducible System: Inducible
Inducing Agent: IPTG
Product Size: 20 µg
Selection Agent (Eukaryotic): None

Contents & storage

20 µg of each pRSET A, B, & C vector is provided lyophilized. TOP10F´ E. coli stab, BL21(DE3)pLysS E. coli stab, and BL21 pRSET/lacZ control E. coli stab are also included. Store E. coli stabs at room temperature. All components are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products