Invitrogen™

Membrane Permeability Dead Cell Apoptosis Kit with PO-PRO™-1 and 7-Aminoactinomycin D, for flow cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This flow cytometry product detects apoptosis on the basis of changes that occur in the permeability of cell membranes using PO-PRO™-1 and 7-aminoactinomycin D (7-AAD) nucleic acid stains. After staining a cell population with PO-PRO™-1 dye and 7-AAD, apoptotic cells show violet fluorescence, dead cells show violet and red fluorescence, and live cells show little or no fluorescence. These populations can easily be distinguished by a flow cytometer that uses both a violet laser and the 488 nm line of an argon-ion laser for excitation. Each kit contains sufficient reagents for approximately 200 flow cytometry tests.

View a selection guide for all apoptosis assays for flow cytometry.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Flow Cytometer Laser Lines: 405, 488
Detection Method: Fluorescent
Excitation⁄Emission (nm): 7-AAD: 546⁄647, PO-PRO™-1: 434⁄456
For Use With (Equipment): Flow Cytometer, Fluorescence Microscope
Format: Slide(s), Tube(s)
Label or Dye: 7-AAD (7-aminoactinomycin D), PO-PRO™-1
Number of Reactions: 200
Product Line: Vybrant®
Product Size: 1 kit
Shipping Condition: Room Temperature

Contents & storage

Contains 1 vial of PO-PRO™-1 dye (500 µL), and 1 vial of 7-AAD (7-aminoactinomycin D, 200 µL).

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products