New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.
Invitrogen™

pRSET-EmGFP Bacterial Expression Vector

Catalog number:  V35320

 Related applications: Protein Assays and Analysis | Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The pRSET-EmGFP, pRSET-CFP and pRSET-BFP vectors allow you to fuse a protein of interest to the widely used and well-characterized fluorescent proteins from the jellyfish Aequorea victoria (1,2) using the pRSET expression vector. These vectors contain the next-generation EGFP, Emerald Green Fluorescent Protein (EmGFP), which has been further optimized for mammalian expression, as well as Cyan (CFP) and Blue (BFP) Fluorescent Proteins for simple, noninvasive detection of recombinant protein.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Vector Type: pRSET
Cloning Method: Restriction Enzyme ⁄ MCS
Promoter: T7
Protein Tag or Fusion: His Tag (6x), Xpress Epitope Tag
Antibiotic Resistance (Bacterial): Ampicillin (AmpR)
Cleavage: EK (Enterokinase) Recognition Site
Constitutive or Inducible System: Inducible
Inducing Agent: IPTG
Product Line: Vivid Colors™
Product Size: 10 µg
Reporter: GFP (EmGFP)
Selection Agent (Eukaryotic): None

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products