New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.

Vivid Colors™ pcDNA™6.2/N-YFP-DEST Vector

Catalog number:  V35820

Invitrogen™  Related applications: Cellular Imaging | Gateway Cloning | Protein Assays and Analysis | Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


The Vivid Colors™ Fluorescent Protein Gateway® Destination Vectors allow you to quickly and easily fuse a protein of interest to the widely used and well-characterized fluorescent proteins from the jellyfish Aequorea victoria (1,2) using the pcDNA™ 6.2 Gateway™ mammalian expression vector. These powerful Gateway® Technology vectors contain the next-generation EGFP, Emerald Green Fluorescent Protein (EmGFP), or the popular Yellow Fluorescent Protein (YFP) for simple, non-invasive detection of recombinant protein. Both fluorescent proteins have been humanized for optimal mammalian expression (3). In addition, the Vivid Colors™ pcDNA™ 6.2 Fluorescent Protein Gateway® Vectors offer:

• CMV promoter for high-level expression of the recombinant fluorescent fusion protein
• Options to fuse EmGFP or YFP to the N- and C-terminus of your protein
For Research Use Only. Not for use in diagnostic procedures.


Promoter: CMV
Key Function: Reporter Assays, Subcellular Localization
Protein Tag or Fusion: V5 Epitope Tag, YFP
Reporter: YFP
Delivery Method: Transfection
Cloning Method: Gateway®
Selection Agent (Eukaryotic): Blasticidin
Vector Type: pDEST, pcDNA
Constitutive or Inducible System: Constitutive
Product Line: Gateway®, Vivid Colors™, pcDNA™
Product Size: 6 µg

Contents & storage

Each vector is supplied as supercoiled and lyophilized plasmid DNA. Store at -20°C after resuspended. Guaranteed stable for 6 months when properly stored.


Manuals & protocols

Recommended products