BLOCK-iT™ Lentiviral RNAi Zeo Gateway™ Vector Kit

Numéro de catalogue:  V48820

Invitrogen™  Applications liées: RNAi

Error lors du chargement de votre contenu!

  Numéro de catalogue
Choisir votre mod�le de prix
Taille unitaire
Prix ({{currency}}) Disponibilité Qté
{{product.sku}} {{product.sku}} également connu comme {{product.formattedSku}} 
{{product.availability.message}}
Ajouts

Votre stock sur site

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} En stock)
›› {{supplyCenter.scName}}(En rupture de stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Haut de la page

Description

The BLOCK-iT™ Lentiviral RNAi Zeo Gateway® Vector Kit contains the pLenti4/BLOCK-iT™-DEST expression vector which enables lentiviral delivery and genomic integration of DNA coding for shRNA. Once expressed, the shRNA is processed by cellular machinery and initiates target-specific RNAi. The pLenti4/BLOCK-iT™-DEST vector offers:

Gateway® Technology for efficient recombination of the RNAi cassette from the BLOCK-iT™ inducible pENTR™/H1/TO vector

• All required components for efficient lentiviral packaging, delivery, and integration of the shRNA
• Zeocin™ selection marker for fast selection of clonal cell lines containing the RNAi cassette
For Research Use Only. Not for use in diagnostic procedures.

Spécifications

Cloning Method: Gateway®
Constitutive or Inducible System: Inducible
Delivery Method: Lentiviral
Product Line: BLOCK-iT™, Gateway®
Product Size: 20 reactions
Promoter: H1⁄TO
RNAi Type: shRNA
Selection Agent (Eukaryotic): Zeocin™
Vector Type: pLenti

Contenu et stockage

The BLOCK-iT™ Lentiviral RNAi Zeo Gateway® Vector Kit contains the pLenti4/BLOCK-iT™-DEST vector, pLenti4-GW/H1/TO-lamin shRNA control vector, and One Shot® Stbl3™ Chemically Competent E.Coli. Store the pLenti4/BLOCK-iT™-DEST, pLenti6/TR vector, Blasticidin, ViraPower™ Lentiviral packaging mix, and the control vector at -20°C. Store the One Shot Stbl3™ Competent Cells 293 FT cell, and LR Clonase® enzyme mix at -80°C. Store Lipofectamine™ 2000 at +4°C. All reagents are guaranteed stable for 6 months when properly stored.

Documents

Manuels et protocoles