pOG44 Flp-Recombinase Expression Vector

Catalog number:  V600520

Invitrogen™  Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

In addition to the original pcDNA™5/FRT vector, four more Flp-In™ expression vectors-pcDNA5/FRT/V5-His-TOPO®, pSecTag/FRT/V5-His-TOPO®, pEF5/FRT/V5-DEST™, and pEF5/FRT/V5-D-TOPO® (Figure 1)-are available for cloning and expressing in the Flp-In™ System. These vectors offer a variety of cloning options and different promoter types. Vectors include the following features:

• Flp Recombinase Target (FRT) site for efficient integration into Flp-In™ Cell Lines
• Hygromycin resistance gene for convenient selection of integrants
• C-terminal V5 tag for easy detection of fusion proteins with the Anti-V5 Antibody
• 6xHis tag (pcDNA5/FRT/V5-His-TOPO® and pSecTag/FRT/V5-His-TOPO® vectors only) for rapid purification of fusion proteins on nickel-chelating resin

All vectors feature either the high-level CMV or EF-1α promoter. In addition pSecTag/FRT/V5-His-TOPO® contains the Igκ leader sequence for secretion of recombinant protein.

Entry Options
pcDNA™5/FRT is suitable for restriction digest-mediated cloning. The other four Flp-In™ expression vectors offer two time-saving options for cloning into a Flp-In™ expression vector:
Five-minute TOPO® Cloning

The pcDNA™5/FRT/V5-His and pSecTag/FRT/V5-His TOPO® TA Expression Kits offer five-minute cloning of Taq-amplified PCR products directly into a Flp-In™ expression vector. The pEF5/FRT/V5 Directional TOPO® Expression Kit allows five-minute directional cloning of PCR product generated with a proofreading enzyme. Using Directional TOPO® Cloning, >90% of the resulting clones will be in the correct orientation for expression, reducing time spent colony screening.
Easy Recombination into a Gateway® vector

The pEF5/FRT/V5-DEST™ vector is compatible with Gateway® Technology* for easy transfer of your gene of interest into different vectors or host systems. The fast, efficient Gateway® recombination reaction can be simultaneously performed with multiple destination vectors, saving you time when working with different systems.

Choice of Promoters
Flp-In™ expression vectors are available with the CMV or EF-1α promoter. The pEF5/FRT/V5-DEST™ and pEF5/FRT/V5-D-TOPO® vectors contain the human EF-1É promoter for driving expression of the gene of interest. The EF-1α promoter expresses in a wide range of mammalian cell types, including those where the CMV promoter expression is absent or inconsistent. This promoter may be more appropriate for long-term gene expression in some cell types and is recommended for expression in the BHK and 3T3 pre-made Flp-In™ Cell Lines. The pcDNA5/FRT/V5-His-TOPO® and pSecTag/FRT/V5-His-TOPO® vectors carry the CMV promoter for high-level constitutive expression of your gene of interest in most cell types.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Promoter: CMV
Key Function: Targeted Integration
Protein Tag or Fusion: Untagged
Delivery Method: Transfection
Selection Agent (Eukaryotic): None
Vector Type: pOG44
Constitutive or Inducible System: Constitutive
Product Line: Flp-In™
Product Size: 20 µg

Contents & storage

The pcDNA™5/FRT/V5-His TOPO® TA Expression Kit, pSecTag/FRT/V5-His TOPO® TA Expression Kit, and the pEF5/FRT/V5 Directional TOPO® Expression Kit contain two boxes. The TOPO® Cloning box contains all of the reagents necessary to clone a Taq-amplified PCR product (TOPO® TA) or a PCR product generated with a proofreading enzyme (Directional TOPO®), including 200 ng of topoisomerase Iactivated vector, sterile water, dNTPs, 10X PCR buffer, salt solution, control template and primers, primers for sequencing, and an expression control plasmid. Store at -20°C. The One Shot® Competent Cell box contains all of the reagents necessary for transformation, including twenty-one 50-µl aliquots of One Shot® TOP10 Chemically Competent E. coli, S.O.C. medium, and pUC19 supercoiled control plasmid. Store at -80°C. The pEF5/FRT/V5-DEST™ vector contains 6 µg of lyophilized vector and a control plasmid. Store at -20°C once resuspended. All reagents are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols