pcDNA™5/FRT Mammalian Expression Vector

Catalog number: V601020

Invitrogen™  Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

pcDNA5⁄FRT vector is a 5.1 kb expression vector designed for use with the Flp-In™ System. This vector features: 1. CMV promoter for high level expression in mammalian cells; 2. ten unique restriction site for easy cloning; 3. FLP Recombination Target (FRT) site for Flp recombinase-mediated integration of the vector into the Flp-In™ host cell line and 4. Hygromycin resistance gene for selection of stable cell lines.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Promoter: CMV
Key Function: Targeted Integration
Protein Tag or Fusion: Untagged
Delivery Method: Transfection
Cloning Method: Restriction Enzyme ⁄ MCS
Selection Agent (Eukaryotic): Hygromycin
Vector Type: pcDNA
Constitutive or Inducible System: Constitutive
Product Line: Flp-In™, pcDNA™
Product Size: 20 µg

Contents & storage

Contains both pcDNA5⁄FRT and pcDNA5⁄FRT⁄CAT vectors. Store at -20°C

Documents

Manuals & protocols

Recommended products