pEF5/FRT/V5-DEST™ Gateway™ Vector

Catalog number:  V602020

Invitrogen™  Related applications: Gateway Cloning | Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


To fit all of your expression needs, Invitrogen offers state-of-the-art Gateway® destination vectors for expression in E. coli, insect, yeast, or mammalian cells, as well as for production of native protein or N- or C-terminal fusion proteins. All Gateway® destination vectors have attR sites for recombination with any attL-flanked fragment, regardless of whether it is an entry clone or an Ultimate™ RF Clone. The following table lists the wide range of destination vectors available.

Additional materials required, available separately: Gateway® entry clone, Gateway® LR Clonase® enzyme mix, and reaction buffer.
For Research Use Only. Not for use in diagnostic procedures.


Promoter: EF-1α
Key Function: Targeted Integration
Protein Tag or Fusion: V5 Epitope Tag
Delivery Method: Transfection
Cloning Method: Gateway®
Selection Agent (Eukaryotic): Hygromycin
Vector Type: pDEST
Constitutive or Inducible System: Constitutive
Product Line: Flp-In™, Gateway®
Product Size: 6 µg

Contents & storage

All destination vectors are provided lyophilized and supercoiled.


Manuals & protocols

Recommended products