pFRT/lacZeo2 Vector

Catalog number:  V602220

Invitrogen™  Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


pFRT/lacZeo and pFRT/lacZeo2 were constructed for establishing Flp-In™ Cell Lines that contain a single FRT site. Each vector includes a:

• FRT site for subsequent targeted integration of a Flp-In™ expression vector
Z-Zeocin™ fusion for selection of stable integrants with Zeocin™

These vectors differ in the SV40 promoter that drives expression of the lacZ-Zeocin™ fusion. pFRT/lacZeo2 contains a truncated version of the SV40 promoter. Use of this vector facilitates the isolation of clones that have integrated the vector near enhancer elements in the genome. Selection of stable clones using pFRT/lacZeo2 may allow easier isolation of clones that express a gene of interest at higher levels.
For Research Use Only. Not for use in diagnostic procedures.


Promoter: SV40
Key Function: Targeted Integration
Protein Tag or Fusion: Untagged
Delivery Method: Transfection
Selection Agent (Eukaryotic): Zeocin™
Vector Type: pFRT
Constitutive or Inducible System: Constitutive
Product Line: Flp-In™
Product Size: 20 µg

Contents & storage

20 µg pFRT/lacZeo2 is supplied at 0.5 µg/µL in 10 mM Tris-HCl, 1 mM EDTA, pH 8.0 in a total volume of 40 µL.


Manuals & protocols