pShooter™ Mammalian Expression Vector (pCMV/myc/ER)

Catalog number:  V82320

Invitrogen™  Related applications: Cell Structure | Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The pShooter™ vectors are designed to localize recombinant proteins to the nucleus, mitochondria, endoplasmic reticulum, or cytoplasm. In addition to the localization signal, each pShooter™ vector contains:

• Strong mammalian promoter (CMV or EF-1É) for constitutive expression of your protein of interest
• C-terminal c-myc epitope for detection with an Anti-myc Antibody
• Bovine Growth Hormone (BGH) polyadenylation site for mRNA stability
• f1 origin for single-stranded rescue of sense DNA
• Neomycin resistance gene for selection of mammalian cells with Geneticin¤ selection agent
• Ampicillin resistance and pUC origin for selection and maintenance in E. coli

Each vector is provided with a positive control plasmid. The positive control expresses GFP and targets it to the same subcellular location as the pShooter™ vector.
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Promoter: CMV
Key Function: Subcellular Localization
Protein Tag or Fusion: c-Myc Epitope Tag
Delivery Method: Transfection
Cloning Method: Restriction Enzyme ⁄ MCS
Selection Agent (Eukaryotic): Geneticin® (G-418)
Vector Type: pCMV
Constitutive or Inducible System: Constitutive
Product Line: pShooter™
Product Size: 20 µg

Contents & storage

20 µg of the pShooter™ vector and GFP control are supplied supercoiled and lyophilized. Store at -20°C. Vectors are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols