pcDNA™4/HisMax A, B, & C Mammalian Expression Vectors

Catalog number:  V86420

Invitrogen™  Related applications: Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

pcDNA™4/HisMax is specifically designed to maximize protein expression in a variety of mammalian cells. The vector contains the QBI SP163 translational enhancer to increase expression levels two- to five-fold above those seen with the promoter alone (Figure 1). In addition to SP163-enhanced expression, pcDNA™4/HisMax includes a cleavable N-terminal Xpress™ tag for rapid detection of recombinant protein with an Anti-Xpress™ Antibody (Figure 2). pcDNA™4/HisMax is available TOPO® Cloning-ready for five-minute cloning of Taqamplified PCR products.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Promoter: CMV
Key Function: Constitutive Expression
Protein Tag or Fusion: His Tag (6x), Xpress Epitope Tag
Delivery Method: Transfection
Cloning Method: Restriction Enzyme ⁄ MCS
Selection Agent (Eukaryotic): Zeocin™
Vector Type: pcDNA
Cleavage: EK (Enterokinase) Recognition Site
Product Line: pcDNA™
Product Size: 20 µg

Contents & storage

The pcDNA™4/HisMax A, B, & C vectors, 20 µg each, are supercoiled and provided lyophilized. 20 µg of pcDNA™4/HisMax/lacZ control plasmid is also provided. Store vectors at -20°C. The pcDNA™4/HisMax TOPO®TA Expression Kit contains two boxes. The TOPO® TA box contains 200 ng of linearized and topoisomerase I-activated pcDNA4/HisMax-TOPO® vector, dNTPs, control PCR template and primers, forward and reverse primers for sequencing and PCR screening, and a lacZ expression control plasmid.
Store this box at -20°C. The One Shot® box contains transformation reagents including twenty-one 50-µl aliquots of TOP10 E. coli, S.O.C. medium, and a pUC19 supercoiled control plasmid. The One Shot® box should be stored at -80°C. Guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols