pTracer™-SV40 Mammalian Expression Vector

Catalog number:  V87120

Invitrogen™  Related applications: Antibody & Cell Culture Production | Cellular Imaging | Protein Assays and Analysis | Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Three pTracer™ mammalian expression vectors are available that express GFP fused to the selectable marker Zeocin™ . Each vector uses a different set of promoters to express the gene of interest and the Cycle 3 GFP-Zeocin™ fusion.

All three pTracer™ vectors have the following features:

• Cycle 3 GFP-Zeocin™ fusion for selection in mammalian cell lines
• BGH polyA signal and transcription termination sequences to enhance mRNA stability

The pTracer™-SV40 vector offers:
• SV40 promoter for high-level constitutive expression of your gene of interest
• CMV promoter for high-level constitutive expression of the Cycle 3 GFP-Zeocin™ fusion
• SV40 origin for episomal replication and simple vector rescue in cell lines expressing the SV40 large T antigen

The pTracer™-CMV2 vector offers:
• CMV promoter for high-level constitutive expression of your gene of interest
• EF-1α promoter for high-level constitutive expression of the Cycle 3 GFP-Zeocin™ fusion
• Ampicillin resistance gene for selection in E. coli

The pTracer™-EF vectors offer:
• EF-1α promoter for high-level constitutive expression of your gene of interest
• CMV promoter for high-level constitutive expression of the Cycle 3 GFP-Zeocin™ fusion
• C-terminal V5 epitope tag for simple detection of recombinant fusion proteins with an Anti-V5 Antibody
• C-terminal 6xHis tag for rapid purification on nickel-chelating resin and simple detection with an Anti-His(C-term) Antibody
• Multiple cloning site supplied in three reading frames to simplify cloning in frame with the C-terminal tag
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Promoter: SV40
Key Function: Reporter Assays
Protein Tag or Fusion: Untagged
Reporter: GFP (Cycle 3)
Delivery Method: Transfection
Cloning Method: Restriction Enzyme ⁄ MCS
Selection Agent (Eukaryotic): Zeocin™
Vector Type: pTracer
Constitutive or Inducible System: Constitutive
Product Line: pTRACER™
Product Size: 20 µg

Contents & storage

pTracer Kits contain 20 µg of the appropriate vector and 1 g of Zeocin (100 mg/ml). Store -20°C. All components for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products